Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HVA22J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129161.3 | NP_565832.1 | 5 - 6 | 455 | 109 | |
GenBank mRNA | DQ446602.1 | - | 3 - 4 | 185 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |