Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202772.1 | NP_001189701.1 | 3 - 4 | 1050 | 78 | |
NM_129396.4 | NP_181374.1 | 3 - 4 | 1096 | 78 | ||
GenBank mRNA | AF166352.1 | - | 3 - 4 | 1022 | 78 | |
AY085348.1 | - | 3 - 4 | 1050 | 78 | ||
BT001918.1 | - | 3 - 4 | 962 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |