Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IREG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336699.1 | NP_001323536.1 | 1 - 2 | 593 | 109 | |
NM_001336700.1 | NP_001323537.1 | 1 - 2 | 593 | 109 | ||
NM_001336701.1 | NP_001323539.1 | 2 - 3 | 436 | 109 | ||
NM_001336702.1 | NP_001323540.1 | 2 - 3 | 404 | 109 | ||
NM_001336703.1 | NP_001323535.1 | 2 - 3 | 460 | 109 | ||
NM_001336704.1 | NP_001323538.1 | 2 - 3 | 385 | 109 | ||
NM_001336705.1 | NP_001323541.1 | 2 - 3 | 292 | 109 | ||
NM_129402.6 | NP_181380.1 | 2 - 3 | 356 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |