Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129637.3 | NP_181607.1 | 1 - 2 | 587 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |