Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129647.4 | NP_181616.3 | 20 - 21 | 2658 | 69 | |
GenBank mRNA | AY081315.1 | - | 20 - 21 | 2449 | 69 | |
BT010364.1 | - | 19 - 20 | 2400 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |