Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336913.1 | NP_001323447.1 | 8 - 9 | 1443 | 70 | |
NM_001336914.1 | NP_001323448.1 | 7 - 8 | 1432 | 70 | ||
NM_129709.5 | NP_181677.1 | 8 - 9 | 1464 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |