Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129716.4 | NP_181684.1 | 4 - 5 | 819 | 86 | |
GenBank mRNA | AF378875.1 | - | 4 - 5 | 539 | 86 | |
AF446861.1 | - | 4 - 5 | 517 | 86 | ||
AJ347732.1 | - | 4 - 5 | 517 | 86 | ||
AY044322.1 | - | 4 - 5 | 517 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |