Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129731.6 | NP_565954.1 | 5 - 6 | 1130 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |