Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129775.3 | NP_181742.2 | 6 - 7 | 715 | 140 | |
NM_201935.3 | NP_973664.1 | 6 - 7 | 917 | 140 | ||
GenBank mRNA | AK221655.1 | - | 6 - 7 | 712 | 140 | |
AY136418.1 | - | 6 - 7 | 712 | 140 | ||
BT008753.1 | - | 6 - 7 | 645 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |