Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036455.1 | NP_001031532.1 | 2 - 3 | 574 | 89 | |
NM_001036456.1 | NP_001031533.1 | 2 - 3 | 574 | 89 | ||
NM_129820.3 | NP_565977.1 | 2 - 3 | 589 | 89 | ||
GenBank mRNA | AB011545.1 | - | 2 - 3 | 475 | 89 | |
AY085927.1 | - | 2 - 3 | 574 | 89 | ||
AY093076.1 | - | 2 - 3 | 572 | 89 | ||
AY128764.1 | - | 2 - 3 | 382 | 89 | ||
U60444.3 | - | 2 - 3 | 441 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |