Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036455.1 | NP_001031532.1 | 6 - 7 | 978 | 83 | |
NM_129820.3 | NP_565977.1 | 6 - 7 | 996 | 83 | ||
GenBank mRNA | AB011545.1 | - | 6 - 7 | 882 | 83 | |
AY085927.1 | - | 6 - 7 | 981 | 83 | ||
AY093076.1 | - | 6 - 7 | 979 | 83 | ||
AY128764.1 | - | 6 - 7 | 789 | 83 | ||
U60444.3 | - | 6 - 7 | 848 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |