Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084574.1 | NP_001078043.1 | 7 - 8 | 1217 | 115 | |
NM_129850.4 | NP_565990.1 | 7 - 8 | 1348 | 115 | ||
NM_201945.3 | NP_973674.1 | 6 - 7 | 1391 | 115 | ||
GenBank mRNA | AF367316.1 | - | 7 - 8 | 1174 | 115 | |
AY143940.1 | - | 7 - 8 | 1090 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |