Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129913.3 | NP_181880.1 | 1 - 2 | 246 | 82 | |
GenBank mRNA | AK175705.1 | - | 1 - 2 | 105 | 82 | |
AK176542.1 | - | 1 - 2 | 106 | 82 | ||
AK176838.1 | - | 1 - 2 | 110 | 82 | ||
AY085600.1 | - | 1 - 2 | 110 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |