Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100303.4 | NP_171918.1 | 1 - 2 | 1076 | 179 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |