Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PP2-B13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104503.5 | NP_176020.1 | 2 - 3 | 531 | 180 | |
GenBank mRNA | BT012215.1 | - | 2 - 3 | 463 | 180 | |
BT015705.1 | - | 2 - 3 | 363 | 180 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |