Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PP2A-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104676.5 | NP_176192.1 | 5 - 6 | 953 | 143 | |
GenBank mRNA | AY063942.1 | - | 5 - 6 | 895 | 143 | |
AY096543.1 | - | 5 - 6 | 743 | 143 | ||
M96732.1 | - | 5 - 6 | 833 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |