Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332792.1 | NP_001322427.1 | 4 - 5 | 1148 | 156 | |
NM_102559.2 | NP_174115.1 | 5 - 6 | 1077 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |