Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332948.1 | NP_001322636.1 | 3 - 4 | 723 | 164 | |
NM_102856.3 | NP_174403.1 | 3 - 4 | 656 | 164 | ||
GenBank mRNA | BT025974.1 | - | 3 - 4 | 623 | 164 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |