Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXLA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114466.1 | NP_190183.1 | 1 - 2 | 195 | 130 | |
GenBank mRNA | AF378896.1 | - | 1 - 2 | 195 | 130 | |
AY052745.1 | - | 1 - 2 | 123 | 130 | ||
AY058142.1 | - | 1 - 2 | 195 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |