Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT76E11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114534.3 | NP_190251.1 | 1 - 2 | 643 | 81 | |
GenBank mRNA | AY080716.1 | - | 1 - 2 | 642 | 81 | |
AY117336.1 | - | 1 - 2 | 473 | 81 | ||
KJ138802.1 | - | 1 - 2 | 473 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |