Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115627.4 | NP_191326.1 | 1 - 2 | 880 | 156 | |
GenBank mRNA | AF254447.1 | - | 1 - 2 | 733 | 156 | |
BT021089.1 | - | 1 - 2 | 713 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |