Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PP2A-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115712.4 | NP_567066.1 | 6 - 7 | 641 | 165 | |
GenBank mRNA | AY056222.1 | - | 6 - 7 | 607 | 165 | |
AY057604.1 | - | 6 - 7 | 591 | 165 | ||
AY113023.1 | - | 6 - 7 | 416 | 165 | ||
U08047.1 | - | 6 - 7 | 535 | 165 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |