Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATPMEPCRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116466.4 | NP_567227.1 | 1 - 2 | 1152 | 136 | |
GenBank mRNA | AY150433.1 | - | 1 - 2 | 1023 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |