Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP79A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342806.1 | NP_001318481.1 | 2 - 3 | 876 | 167 | |
NM_120608.2 | NP_568153.2 | 1 - 2 | 980 | 167 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |