Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123591.2 | NP_199041.1 | 1 | 865 | 150 | |
GenBank mRNA | DQ056704.1 | - | 1 | 865 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |