Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVA-P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342299.1 | NP_001329512.1 | 3 | 1059 | 127 | |
NM_119638.3 | NP_195198.1 | 3 | 794 | 127 |
Gene Symbol: AT2G16510 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF372872.1 | - | 3 | 688 | 127 | |
AY086410.1 | - | 3 | 756 | 127 | ||
L44581.1 | - | 3 | 671 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |