Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035884.2 | NP_001030961.1 | 1 | 1092 | 76 | |
NM_100343.3 | NP_171958.1 | 2 | 797 | 76 | ||
GenBank mRNA | AB020975.1 | - | 2 | 740 | 76 | |
AF428288.1 | - | 2 | 711 | 76 | ||
AX765842.1 | - | 1 | 661 | 76 | ||
AY133628.1 | - | 1 | 661 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |