Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036114.1 | NP_001031191.1 | 2 - 3 | 245 | 146 | |
NM_104340.5 | NP_564663.1 | 2 - 3 | 407 | 146 | ||
GenBank mRNA | AY054604.1 | - | 2 - 3 | 233 | 146 | |
AY072527.1 | - | 2 - 3 | 160 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |