Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128510.3 | NP_180517.1 | 3 - 4 | 724 | 134 | |
GenBank mRNA | BT029466.1 | - | 3 - 4 | 569 | 134 | |
EU072918.1 | - | 3 - 4 | 569 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |