Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTFL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084326.2 | NP_001077795.1 | 1 | 214 | 103 | |
NM_001160987.1 | NP_001154459.1 | 2 | 244 | 103 | ||
GenBank mRNA | AY254321.1 | - | 1 | 73 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |