Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075135.1 | NP_001068603.1 | 2 - 3 | 276 | 70 | |
GenBank mRNA | AB099076.1 | - | 2 - 3 | 226 | 70 | |
BC102425.1 | - | 2 - 3 | 276 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |