Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKN2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101054.2 | NP_001094524.1 | 1 - 2 | 244 | 114 | |
GenBank mRNA | BC151477.1 | - | 1 - 2 | 245 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |