Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001139.2 | NP_001001139.1 | 1 - 2 | 397 | 88 | |
GenBank mRNA | X55010.1 | - | 398 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |