Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007811.2 | NP_001007812.2 | 16 - 17 | 1848 | 68 | |
GenBank mRNA | AY788896.1 | - | 7 - 8 | 768 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |