Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFAIP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007813.2 | NP_001007814.1 | 2 - 3 | 301 | 79 | |
GenBank mRNA | AJ854251.1 | - | 2 - 3 | 303 | 79 | |
BC151789.1 | - | 2 - 3 | 315 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |