Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008416.1 | NP_001008416.1 | 2 - 3 | 250 | 75 | |
GenBank mRNA | AY290705.1 | - | 2 - 3 | 250 | 75 | |
BC151558.1 | - | 2 - 3 | 245 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |