Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSENEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008669.1 | NP_001008669.1 | 2 - 3 | 208 | 81 | |
GenBank mRNA | AY821679.1 | - | 2 - 3 | 208 | 81 | |
BC102375.1 | - | 2 - 3 | 290 | 81 | ||
MK424003.1 | - | 1 - 2 | 145 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |