Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011683.2 | NP_001011683.1 | 4 - 5 | 1797 | 149 | |
GenBank mRNA | AY514443.1 | - | 4 - 5 | 1798 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Bos taurus Inventoried Probe spans exons |