Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014387.2 | NP_001014409.1 | 3 - 4 | 275 | 80 | |
GenBank mRNA | AB098828.1 | - | 2 - 3 | 263 | 80 | |
AB099081.1 | - | 4 - 5 | 304 | 80 | ||
AY528251.1 | - | 3 - 4 | 277 | 80 | ||
AY911345.1 | - | 4 - 5 | 320 | 80 | ||
BC102500.1 | - | 1 | 321 | 80 | ||
DQ347612.1 | - | 4 - 5 | 321 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |