Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032299.1 | NP_001027470.1 | 4 - 5 | 577 | 127 | |
GenBank mRNA | AF450088.1 | - | 2 - 3 | 184 | 127 | |
AJ699418.1 | - | 4 - 5 | 577 | 127 | ||
BC149573.1 | - | 4 - 5 | 1041 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |