Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173903.4 | NP_776328.2 | 2 - 3 | 745 | 63 | |
GenBank mRNA | BC142181.1 | - | 2 - 3 | 611 | 63 | |
DV831465.1 | - | 2 - 3 | 369 | 63 | ||
X04411.1 | - | 376 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |