Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173908.2 | NP_776333.1 | 4 - 5 | 511 | 107 | |
GenBank mRNA | BC103184.1 | - | 4 - 5 | 510 | 107 | |
X95310.1 | - | 511 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |