Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173980.2 | NP_776405.1 | 4 - 5 | 368 | 77 | |
GenBank mRNA | AF541976.1 | - | 4 - 5 | 368 | 77 | |
AJ011680.1 | - | 4 - 5 | 368 | 77 | ||
BC102211.1 | - | 4 - 5 | 390 | 77 | ||
BT029909.1 | - | 4 - 5 | 378 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |