Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174006.2 | NP_776431.1 | 2 - 3 | 243 | 92 | |
GenBank mRNA | EU276059.1 | - | 2 - 3 | 217 | 92 | |
M84602.1 | - | 2 - 3 | 243 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Bos taurus Inventoried Probe spans exons |