Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICAM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174349.1 | NP_776774.1 | 4 - 5 | 1033 | 109 | |
GenBank mRNA | L41844.1 | - | 4 - 5 | 1033 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |