Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174536.2 | NP_776961.1 | 6 - 7 | 880 | 135 | |
GenBank mRNA | AF005940.1 | - | 6 - 7 | 880 | 135 | |
BC109518.1 | - | 6 - 7 | 991 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |