Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174544.2 | NP_776969.1 | 6 - 7 | 730 | 140 | |
GenBank mRNA | X05718.1 | - | 730 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Bos taurus Probe spans exons |