Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174802.2 | NP_777227.1 | 6 - 7 | 711 | 57 | |
GenBank mRNA | U79414.1 | - | 6 - 7 | 711 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |