Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174830.1 | NP_777255.1 | 4 - 5 | 391 | 84 | |
GenBank mRNA | AF141872.1 | - | 4 - 5 | 391 | 84 | |
BC109922.1 | - | 4 - 5 | 396 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |