Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176646.3 | NP_788819.1 | 2 - 3 | 679 | 75 | |
GenBank mRNA | AB026444.1 | - | 2 - 3 | 680 | 75 | |
BC109553.1 | - | 2 - 3 | 697 | 75 | ||
BT030731.1 | - | 2 - 3 | 682 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |